Learn More
Biomatik Corporation Human Transmembrane Protein 2 Like Protein (TMEM2L) ELISA Kit, 96 tests
Catalog No. 501504015
Encompass_Preferred
Human Transmembrane Protein 2 Like Protein (TMEM2L) ELISA Kit for detecting TMEM2L in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8WUJ3. Gene ID: 57214. Target Synonyms: CEMIP; CCSP1; KIAA1199; Cell Migration Inducing Protein, Hyaluronan Binding. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4015 Supplier Biomatik Corporation Supplier No. EKU07858
Human Transmembrane Protein 2 Like Protein (TMEM2L) ELISA Kit for detecting TMEM2L in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8WUJ3. Gene ID: 57214. Target Synonyms: CEMIP; CCSP1; KIAA1199; Cell Migration Inducing Protein, Hyaluronan Binding. Restrictions: For Research Use Only. Not for use in diagnostic procedures.