Learn More
Biomatik Corporation Carboxymethyl Lysine (CML) ELISA Kit
Supplier:  Biomatik Corporation EKL61716
Encompass_Preferred
Carboxymethyl Lysine (CML) ELISA Kit for detecting CML in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Universal. Detection Range: 78.1-5000ng/ml. Sensitivity: 33ng/ml. Format: 96-Well Plate. Assay Type: Competitive inhibition. Assay Time: 3h. Target Synonyms: N(6)-Carboxymethyllysine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8829