Learn More
Biomatik Corporation Human Transthyretin (TTR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07864
Encompass_Preferred
Human Prealbumin (PALB) ELISA Kit for detecting PALB in serum, plasma, tissue homogenates, cell lysates, cerebrospinal fluid, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.151ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02766. Gene ID: 7276. Target Synonyms: TBPA; PA; PLB; ATTR; TTR; Transthyretin; Prealbumin Amyloidosis Type I; Transports Thyroxine and Retinol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4016