Learn More
Biomatik Corporation Mouse Follicle Stimulating Hormone (FSH) ELISA Kit
Supplier:  Biomatik Corporation EKL61725
Encompass_Preferred
Mouse Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.49ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: Q60687. Gene ID: 14308. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8838