Learn More
Biomatik Corporation Mouse Trefoil Factor 2 (TFF2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07872
Encompass_Preferred
Mouse Trefoil Factor 2 (TFF2) ELISA Kit for detecting TFF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q03404. Target Synonyms: SML1; SP; Spasmolytic Protein 1; Spasmolysin; Spasmolytic polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4020