Learn More
Biomatik Corporation Mouse Trefoil Factor 3, Intestinal (TFF3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07874
Encompass_Preferred
Mouse Trefoil Factor 3 (TFF3) ELISA Kit for detecting TFF3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62395. Gene ID: 21786. Target Synonyms: HITF; ITF3; TFI; HP1.B; Trefoil Factor 3, Intestinal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4022