Learn More
Biomatik Corporation Human Triggering Receptor Expressed On Myeloid Cells 1 (TREM1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07879
Encompass_Preferred
Human Triggering Receptor Expressed On Myeloid Cells 1 (TREM1) ELISA Kit for detecting TREM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NP99. Gene ID: 54210. Target Synonyms: CD354; Triggering Receptor Expressed On Monocytes-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4023