Learn More
Biomatik Corporation Human Triosephosphate Isomerase 1 (TPI1) ELISA Kit, 96 tests
Catalog No. 501504026
Encompass_Preferred
Human Triosephosphate Isomerase 1 (TPI1) ELISA Kit for detecting TPI1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.269ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60174. Gene ID: 7167. Target Synonyms: TIM; Triose-phosphate isomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4026 Supplier Biomatik Corporation Supplier No. EKU07885
Human Triosephosphate Isomerase 1 (TPI1) ELISA Kit for detecting TPI1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.269ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60174. Gene ID: 7167. Target Synonyms: TIM; Triose-phosphate isomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.