Learn More
Biomatik Corporation Human Tripeptidyl Peptidase I (TPP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07887
Encompass_Preferred
Human Tripeptidyl Peptidase I (TPP1) ELISA Kit for detecting TPP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14773. Gene ID: 1200. Target Synonyms: CLN2; GIG1; LPIC; Tripeptidyl aminopeptidase; Lysosomal pepstatin-insensitive protease; Ceroid-Lipofuscinosis,Neuronal 2,Late Infantile; Cell growth-inhibiting gene 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4027