Learn More
Biomatik Corporation Human Troponin I Type 3, Cardiac (TNNI3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07900
Encompass_Preferred
Human Cardiac Troponin I (cTnI) ELISA Kit for detecting cTnI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19429. Gene ID: 7137. Target Synonyms: TNNI3; TNNC1; CMH7; C-TnI; Troponin I Type 3; Troponin I Type 3, Cardiac. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4028