Learn More
Biomatik Corporation Mouse Troponin T Type 1, Slow Skeletal (TNNT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07907
Encompass_Preferred
Mouse Troponin T Type 1, Slow Skeletal (TNNT1) ELISA Kit for detecting TNNT1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88346. Gene ID: 21955. Target Synonyms: ANM; STNT; TNT; TNTS; Slow Skeletal Muscle Troponin T. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4033