Learn More
Biomatik Corporation Rat Troponin T Type 2, Cardiac (TNNT2) ELISA Kit, 96 tests
Catalog No. 501504036
Encompass_Preferred
Rat Troponin T Type 2, Cardiac (TNNT2) ELISA Kit for detecting TNNT2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50753. Gene ID: 24837. Target Synonyms: CMH2; cTnT; TnTc; Cardiomyopathy,Hypertrophic 2; Cardiac muscle troponin T. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4036 Supplier Biomatik Corporation Supplier No. EKU07912
Rat Troponin T Type 2, Cardiac (TNNT2) ELISA Kit for detecting TNNT2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50753. Gene ID: 24837. Target Synonyms: CMH2; cTnT; TnTc; Cardiomyopathy,Hypertrophic 2; Cardiac muscle troponin T. Restrictions: For Research Use Only. Not for use in diagnostic procedures.