Learn More
Biomatik Corporation Rat Tryptase (TPS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07925
Encompass_Preferred
Rat Tryptase (TPS) ELISA Kit for detecting TPS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27435. Gene ID: 54271. Target Synonyms: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4040