Learn More
Biomatik Corporation Human Tryptase Beta 2 (TPSb2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07926
Encompass_Preferred
Human Tryptase Beta 2 (TPSb2) ELISA Kit for detecting TPSb2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 2.43pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20231. Gene ID: 64499. Target Synonyms: TPS2; Tryptase Beta II; Tryptase Beta III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4041