Learn More
Biomatik Corporation Human Tubulin Polymerization Promoting Protein (TPPP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07938
Encompass_Preferred
Human Tubulin Polymerization Promoting Protein (TPPP) ELISA Kit for detecting TPPP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O94811. Gene ID: 11076. Target Synonyms: TPPP1; p25alpha; p25; TPPP/p25; Brain Specific Protein p25 Alpha; 25 kDa brain-specific protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4044