Learn More
Biomatik Corporation Dog Tumor Necrosis Factor Alpha (TNFa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07942
Encompass_Preferred
The Dog Tumor Necrosis Factor Alpha (TNFa) ELISA Kit allows for quantitative determination of Tumor Necrosis Factor Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51742. Cross Activity: No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4046