Learn More
Biomatik Corporation Guinea Pig Tumor Necrosis Factor Alpha (TNFa) ELISA Kit, 96 tests
Catalog No. 501504047
Encompass_Preferred
The Guinea Pig Tumor Necrosis Factor Alpha (TNFa) ELISA Kit allows for quantitative determination of Tumor Necrosis Factor Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Cavia (Guinea pig). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51435. Cross Activity: No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4047 Supplier Biomatik Corporation Supplier No. EKU07945
The Guinea Pig Tumor Necrosis Factor Alpha (TNFa) ELISA Kit allows for quantitative determination of Tumor Necrosis Factor Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Cavia (Guinea pig). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51435. Cross Activity: No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha and analogues was observed. Restriction: For Research Use Only.