Learn More
Biomatik Corporation Rat Tumor Necrosis Factor Alpha (TNFa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07950
Encompass_Preferred
Rat Tumor Necrosis Factor Alpha (TNFa) ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16599. Gene ID: 24835. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4052