Learn More
Biomatik Corporation Rabbit Tumor Necrosis Factor Beta (TNFb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07960
Encompass_Preferred
Rabbit Tumor Necrosis Factor Beta (TNFb) ELISA Kit for detecting TNFb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10154. Gene ID: 100328931. Target Synonyms: LT; LTA; TNF-B; TNFSF1; Tumor Necrosis Factor Ligand Superfamily Member 1; Lymphotoxin Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4056