Learn More
Biomatik Corporation Human Tumor Necrosis Factor Ligand Superfamily, Member 7 (TNFSF7) ELISA Kit, 96 tests
Catalog No. 501504061
Encompass_Preferred
Human Tumor Necrosis Factor Ligand Superfamily, Member 7 (TNFSF7) ELISA Kit for detecting TNFSF7 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32970. Gene ID: 970. Target Synonyms: CD70; CD27L; CD27LG; TNFSF7; CD27 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4061 Supplier Biomatik Corporation Supplier No. EKU07969
Human Tumor Necrosis Factor Ligand Superfamily, Member 7 (TNFSF7) ELISA Kit for detecting TNFSF7 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32970. Gene ID: 970. Target Synonyms: CD70; CD27L; CD27LG; TNFSF7; CD27 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.