Learn More
Biomatik Corporation Human Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) ELISA Kit, 96 tests
Catalog No. 501504064
Encompass_Preferred
Human Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) ELISA Kit for detecting TRAIL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50591. Gene ID: 8743. Target Synonyms: CD253; TNFSF10; APO2L; Apo2-L; TL2; TRAIL; Tumor Necrosis Factor Ligand Superfamily Member 10; Apo-2 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4064 Supplier Biomatik Corporation Supplier No. EKU07981
Human Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) ELISA Kit for detecting TRAIL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50591. Gene ID: 8743. Target Synonyms: CD253; TNFSF10; APO2L; Apo2-L; TL2; TRAIL; Tumor Necrosis Factor Ligand Superfamily Member 10; Apo-2 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.