Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) ELISA Kit, 96 tests
Catalog No. 501504065
Encompass_Preferred
Mouse Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) ELISA Kit for detecting TRAIL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50592. Gene ID: 22035. Target Synonyms: CD253; TNFSF10; APO2L; Apo2-L; TL2; TRAIL; Tumor Necrosis Factor Ligand Superfamily Member 10; Apo-2 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4065 Supplier Biomatik Corporation Supplier No. EKU07982
Mouse Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) ELISA Kit for detecting TRAIL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50592. Gene ID: 22035. Target Synonyms: CD253; TNFSF10; APO2L; Apo2-L; TL2; TRAIL; Tumor Necrosis Factor Ligand Superfamily Member 10; Apo-2 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.