Learn More
Biomatik Corporation Mevalonate Kinase Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50803
Encompass_Preferred
Mevalonate Kinase Colorimetric Cell-Based ELISA for detecting Mevalonate Kinase in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q03426. Gene ID: 4598. Target Synonyms: Mevalonate kinase; MK; MVK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8988