Learn More
Biomatik Corporation Rat Tumor Protein p53 (TP53) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07986
Encompass_Preferred
Rat Tumor Protein p53 (P53) ELISA Kit for detecting P53 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10361. Gene ID: 24842. Target Synonyms: TP53; LFS1; TRP53; Li-Fraumeni Syndrome; Cellular tumor antigen p53; Antigen NY-CO-13; Phosphoprotein p53; Tumor suppressor p53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4067