Learn More
Biomatik Corporation Human Tumor Protein, Translationally Controlled 1 (TPT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07990
Encompass_Preferred
Human Tumor Protein, Translationally Controlled 1 (TPT1) ELISA Kit for detecting TPT1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13693. Gene ID: 7178. Target Synonyms: HRF; TCTP; p02; p23; Fortilin; Translationally-Controlled Tumor Protein; Histamine-releasing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4069