Learn More
Biomatik Corporation Human Tyrosine Hydroxylase (TH) ELISA Kit, 96 tests
Catalog No. 501504072
Encompass_Preferred
Human Tyrosine Hydroxylase (TH) ELISA Kit for detecting TH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07101. Gene ID: 7054. Target Synonyms: TYH; DYT5b; Tyrosine 3-Monooxygenase; Dystonia 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4072 Supplier Biomatik Corporation Supplier No. EKU08002
Human Tyrosine Hydroxylase (TH) ELISA Kit for detecting TH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07101. Gene ID: 7054. Target Synonyms: TYH; DYT5b; Tyrosine 3-Monooxygenase; Dystonia 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.