Learn More
Biomatik Corporation Carbonic Anhydrase IX (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52016
Encompass_Preferred
Carbonic Anhydrase IX (Human) Sandwich ELISA Kit for detecting Carbonic Anhydrase IX in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 32-2000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16790. Gene ID: 768. Target Synonyms: Carbonic anhydrase 9; Carbonate dehydratase IX; CAIX; Membrane antigen MN; P54/58N; Renal cell carcinoma-associated antigen G250; pMW1; G250; MN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9165