Learn More
Biomatik Corporation Human Tyrosine Kinase 2 (Tyk2) ELISA Kit, 96 tests
Catalog No. 501504075
Encompass_Preferred
Human Tyrosine Kinase 2 (Tyk2) ELISA Kit for detecting Tyk2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29597. Gene ID: 7297. Target Synonyms: JTK1; Non-Receptor Tyrosine-Protein Kinase TYK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4075 Supplier Biomatik Corporation Supplier No. EKU08005
Human Tyrosine Kinase 2 (Tyk2) ELISA Kit for detecting Tyk2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29597. Gene ID: 7297. Target Synonyms: JTK1; Non-Receptor Tyrosine-Protein Kinase TYK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.