Learn More
Biomatik Corporation GDF-15 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52044
Encompass_Preferred
GDF-15 (Human) Sandwich ELISA Kit for detecting GDF-15 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99988. Gene ID: 9518. Target Synonyms: Growth/differentiation factor 15; Macrophage inhibitory cytokine 1; MIC-1; NSAID-activated gene 1 protein; Placental TGF-beta; Placental bone morphogenetic protein; Prostate differentiation factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9172