Learn More
Biomatik Corporation Human melanin ELISA kit
Supplier:  Biomatik Corporation EKC34570
Encompass_Preferred
Sample Type: serum. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5 pg/mL-1000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-149-8064