Learn More
Biomatik Corporation 17-Hydroxyprogesterone (17-OHP) ELISA Kit
Supplier:  Biomatik Corporation EKU02011
Encompass_Preferred
17-Hydroxyprogesterone (17-OHP) ELISA Kit for detecting 17-OHP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 1.23-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: 17OHP; 17-OH Progesterone; 17-Alpha-Hydroxyprogesterone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5748