Learn More
Biomatik Corporation Human Ubiquitin Protein Ligase E3A (UBE3A) ELISA Kit, 96 tests
Catalog No. 501504079
Encompass_Preferred
Human Ubiquitin Protein Ligase E3A (UBE3A) ELISA Kit for detecting UBE3A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05086. Gene ID: 7337. Target Synonyms: ANCR; AS; E6-AP; EPVE6AP; HPVE6A; Papilloma Virus E6-Associated,Angelman Syndrome; Oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4079 Supplier Biomatik Corporation Supplier No. EKU08024
Human Ubiquitin Protein Ligase E3A (UBE3A) ELISA Kit for detecting UBE3A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05086. Gene ID: 7337. Target Synonyms: ANCR; AS; E6-AP; EPVE6AP; HPVE6A; Papilloma Virus E6-Associated,Angelman Syndrome; Oncogenic protein-associated protein E6-AP; Renal carcinoma antigen NY-REN-54. Restrictions: For Research Use Only. Not for use in diagnostic procedures.