Learn More
Biomatik Corporation Mouse Uncoupling Protein 1, Mitochondrial (UCP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08037
Encompass_Preferred
Mouse Uncoupling Protein 1, Mitochondrial (UCP1) ELISA Kit for detecting UCP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12242. Gene ID: 22227. Target Synonyms: SLC25A7; UCP; Thermogenin; Solute carrier family 25 member 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4080