Learn More
Biomatik Corporation Human 5-Hydroxytryptamine Receptor 2A (HTR2A) ELISA Kit
Supplier:  Biomatik Corporation EKU02029
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.065ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: 5-HT2A; HTR2; Serotonin Receptor 2A. Restriction: For Research Use Only.
Catalog No. 50-228-5754