Learn More
Biomatik Corporation Fish Cortisol ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31067
Encompass_Preferred
The Fish Cortisol ELISA Kit allows for quantitative determination of Cortisol in serum, plasma, tissue homogenates. Species Reactivity: Fish. Detection Range: 0.0023 ng/mL-10 ng/mL. Sensitivity: 0.0023 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7394