Learn More
Biomatik Corporation Aldosterone (ALD) ELISA Kit
Supplier:  Biomatik Corporation EKU02237
Encompass_Preferred
Aldosterone (ALD) ELISA Kit for detecting ALD in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.41pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5775