Learn More
Biomatik Corporation Mouse Amyloid Beta Peptide 1-40 (Ab1-40) ELISA Kit
Supplier:  Biomatik Corporation EKU02328
Encompass_Preferred
Mouse Amyloid Beta Peptide 1-40 (Ab1-40) ELISA Kit for detecting Ab1-40 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.38pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P12023. Gene ID: 11820. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5786