Learn More
Biomatik Corporation Rat Uromodulin (UMOD) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08058
Encompass_Preferred
Rat Uromodulin (UMOD) ELISA Kit for detecting UMOD in serum, plasma, urine, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27590. Gene ID: 25128. Target Synonyms: THP; Uromucoid,Tamm-Horsfall Glycoprotein; Tamm-Horsfall urinary glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4087