Learn More
Biomatik Corporation Human Uroplakin 3A (UPK3A) ELISA Kit, 96 tests
Catalog No. 501504088
Encompass_Preferred
Human Uroplakin 3A (UPK3A) ELISA Kit for detecting UPK3A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75631. Gene ID: 7380. Target Synonyms: UPIII; UPK3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4088 Supplier Biomatik Corporation Supplier No. EKU08059
Human Uroplakin 3A (UPK3A) ELISA Kit for detecting UPK3A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75631. Gene ID: 7380. Target Synonyms: UPIII; UPK3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.