Learn More
Biomatik Corporation Human Heparin/Platelet Factor 4 (Anti-HPF4) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU02452
Encompass_Preferred
Human Heparin/Platelet Factor 4 (Anti-HPF4) Anti-antigen ELISA Kit for detecting Anti-HPF4 in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P02776. Gene ID: 5196. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5806