Learn More
Biomatik Corporation Human Apelin 13 (AP13) ELISA Kit
Supplier:  Biomatik Corporation EKU02473
Encompass_Preferred
Human Apelin 13 (AP13) ELISA Kit for detecting AP13 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.83pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9ULZ1. Gene ID: 8862. Target Synonyms: APLN13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5809