Learn More
Biomatik Corporation Rat Vanin 1 (VNN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08073
Encompass_Preferred
Rat Vanin 1 (VNN1) ELISA Kit for detecting VNN1 in serum, plasma, urine, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4KLZ0. Gene ID: 29142. Target Synonyms: Pantetheinase; Pantetheine hydrolase; Tiff66; Vascular non-inflammatory molecule 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4092