Learn More
Biomatik Corporation Human Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit, 96 tests
Catalog No. 501504093
Encompass_Preferred
Human Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit for detecting VCAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19320. Gene ID: 7412. Target Synonyms: CD106; INCAM-100; L1CAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4093 Supplier Biomatik Corporation Supplier No. EKU08076
Human Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit for detecting VCAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19320. Gene ID: 7412. Target Synonyms: CD106; INCAM-100; L1CAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.