Learn More
Biomatik Corporation Pig Relaxin (RLN) ELISA Kit
Supplier:  Biomatik Corporation EKU07062
Encompass_Preferred
Pig Relaxin (RLN) ELISA Kit for detecting RLN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.59pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01348. Gene ID: 396891. Target Synonyms: RLXH1; Relaxin H1; Prorelaxin H1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6355