Learn More
Biomatik Corporation Rat Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08079
Encompass_Preferred
Rat Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit for detecting VCAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29534. Gene ID: 25361. Target Synonyms: CD106; INCAM-100; L1CAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4095