Learn More
Biomatik Corporation Rabbit Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08080
Encompass_Preferred
Rabbit Vascular Cell Adhesion Molecule 1 (VCAM1) ELISA Kit for detecting VCAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q865F2. Gene ID: 100008901. Target Synonyms: CD106; INCAM-100; L1CAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4096