Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor 121 (VEGF121) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08082
Encompass_Preferred
Human Vascular Endothelial Growth Factor 121 (VEGF121) ELISA Kit for detecting VEGF121 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15692. Gene ID: 7422. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4097