Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor 165 (VEGF165) ELISA Kit, 96 tests
Catalog No. 501504098
Encompass_Preferred
Human Vascular Endothelial Growth Factor 165 (VEGF165) ELISA Kit for detecting VEGF165 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15692. Gene ID: 7422. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4098 Supplier Biomatik Corporation Supplier No. EKU08086
Human Vascular Endothelial Growth Factor 165 (VEGF165) ELISA Kit for detecting VEGF165 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15692. Gene ID: 7422. Restrictions: For Research Use Only. Not for use in diagnostic procedures.