Learn More
Biomatik Corporation Mouse Vascular Endothelial Growth Factor A (VEGFA) ELISA Kit, 96 tests
Catalog No. 501504101
Encompass_Preferred
Mouse Vascular Endothelial Growth Factor A (VEGFA) ELISA Kit for detecting VEGFA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00731. Gene ID: 22339. Target Synonyms: VEGF; VEGF-A; VPF; Vascular Endothelial Cell Growth Factor A; Vascular Permeability Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4101 Supplier Biomatik Corporation Supplier No. EKU08091
Mouse Vascular Endothelial Growth Factor A (VEGFA) ELISA Kit for detecting VEGFA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00731. Gene ID: 22339. Target Synonyms: VEGF; VEGF-A; VPF; Vascular Endothelial Cell Growth Factor A; Vascular Permeability Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.