Learn More
Biomatik Corporation Mouse Spondin 2 (SPON2) ELISA Kit
Supplier:  Biomatik Corporation EKU07446
Encompass_Preferred
Mouse Spondin 2 (SPON2) ELISA Kit for detecting SPON2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BMS2. Gene ID: 100689. Target Synonyms: DIL1; Mindin; M-spondin; Extracellular Matrix Protein; Differentially expressed in cancerous and non-cancerous lung cells 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6391